Learn More
ASAH1, Mouse, Clone: 1A7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ASAH1.
Supplier: Abnova Corporation H00000427M02
Description
This gene encodes a heterodimeric protein consisting of a nonglycosylated alpha subunit and a glycosylated beta subunit that is cleaved to the mature enzyme posttranslationally. The encoded protein catalyzes the synthesis and degradation of ceramide into sphingosine and fatty acid. Mutations in this gene have been associated with a lysosomal storage disorder known as Farber disease. Multiple transcript variants encoding several distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: PPWTEDCRKSTYPPSGPTYRGAVPWYTINLDLPPYKRWHELMLDKAPMLKVIVNSLKNMINTFVPSGKVMQVVDEKLPGLLGNFPGPFEEEMKGIAAVTDSpecifications
ASAH1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_177924 | |
ASAH1 | |
ASAH1 (NP_808592, 25 a.a. ∼ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG3 κ |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
1A7 | |
Mouse monoclonal antibody raised against a partial recombinant ASAH1. | |
ASAH1 | |
AC/ASAH/FLJ21558/FLJ22079/PHP/PHP32 | |
Mouse | |
Affinity chromatography | |
RUO | |
427 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.