Learn More
ASAP3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ASAP3 peptide using ARM Technology.
Supplier: Abnova Corporation H00055616K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| ASAP3 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| ASAP3 | |
| ASAP3 | |
| A synthetic peptide of human ASAP3 | |
| RUO | |
| 55616 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 | |
| ASAP3 | |
| ACAP4, CENTB6, DDEFL1, FLJ20199, UPLC1 | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.