Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ASC Rabbit anti-Human, Polyclonal

Catalog No. AB3607MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3607MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3607MI Supplier MilliporeSigma™ Supplier No. AB3607
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-ASC Antibody is validated for use in western blotting for the detection of ASC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASC
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Gene Symbols PYCARD; TMS1; MGC10332; ASC; hASC; CARD5
Host Species Rabbit
Immunogen Peptide corresponding to aa 182-195 (RESQSYLVEDLERS) of human ASC.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_145183.1; NM_013258.3; NM_145182.1
Target Species Human
Content And Storage 4°C for 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.