Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASC/TMS1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP178978
Description
ASC/TMS1 Polyclonal specifically detects ASC/TMS1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Western Blot (Negative).Specifications
ASC/TMS1 | |
Polyclonal | |
Unconjugated | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1.12 mg/mL | |
Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:40-1:100, Immunohistochemistry-Paraffin 1:200, Western Blot (Negative) | |
Q9ULZ3 | |
PYCARD | |
This ASC/TMS1 Antibody was developed against a synthetic peptide made to a C-terminal portion of the human ASC/TMS1 protein (between residues 145-195) [Uniprot: Q9ULZ3] | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
29108 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction