Learn More
achaete-scute complex homolog 1 (Drosophila), Mouse, Clone: 3D3, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a member of the basic helix-loop-helix (BHLH) family of transcription factors. The protein activates transcription by binding to the E box (5'-CANNTG-3'). Dimerization with other BHLH proteins is required for efficient DNA binding. This protein plays a role in the neuronal commitment and differentiation and in the generation of olfactory and autonomic neurons. Mutations in this gene may contribute to the congenital central hypoventilation syndrome (CCHS) phenotype in rare cases. [provided by RefSeq]
Specifications
Specifications
| Antigen | achaete-scute complex homolog 1 (Drosophila) |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3D3 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant ASCL1. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ASCL1 |
| Gene Accession No. | NM_004316 |
| Gene Alias | ASH1/HASH1/MASH1/bHLHa46 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.