Learn More
ASCL1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ASCL1 peptide using ARM Technology.
Supplier: Abnova Corporation H00000429K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of the basic helix-loop-helix (BHLH) family of transcription factors. The protein activates transcription by binding to the E box (5'-CANNTG-3'). Dimerization with other BHLH proteins is required for efficient DNA binding. This protein plays a role in the neuronal commitment and differentiation and in the generation of olfactory and autonomic neurons. Mutations in this gene may contribute to the congenital central hypoventilation syndrome (CCHS) phenotype in rare cases. [provided by RefSeq]
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ASCL1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ASCL1 | |
ASCL1 | |
A synthetic peptide of human ASCL1 | |
RUO | |
429 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
achaete-scute complex homolog 1 (Drosophila) | |
ASCL1 | |
ASH1, HASH1, MASH1, bHLHa46 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.