Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASF1a Mouse anti-Human, Clone: MPH7, MilliporeSigma™

Catalog No. MABE90MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE90MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE90MI Supplier MilliporeSigma Supplier No. MABE90

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Use Anti-ASF1a Antibody, clone MPH7 (Mouse Monoclonal Antibody) validated in immunoprecipitation to detect ASF1a also known as Histone chaperone ASF1A, hAsf1 or Asf1a.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASF1a
Applications Immunoprecipitation
Classification Monoclonal
Clone MPH7
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_054753
Gene Symbols ASF1A, CGI-98, HSPC146
Host Species Mouse
Immunogen GST tagged recombinant protein corresponding to human ASF1a.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.