Learn More
ASF1A, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ASF1A peptide using ARM Technology.
Supplier: Abnova Corporation H00025842K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The protein is a key component of a histone donor complex that functions in nucleosome assembly. It interacts with histones H3 and H4, and functions together with a chromatin assembly factor during DNA replication and repair. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ASF1A | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ASF1A | |
ASF1A | |
A synthetic peptide of human ASF1A | |
RUO | |
25842 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ASF1 anti-silencing function 1 homolog A (S. cerevisiae) | |
ASF1A | |
CGI-98, CIA, DKFZp547E2110, HSPC146 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.