Learn More
ASGPR 1 Mouse, Unlabeled, Clone: 8D7, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 563654
Description
The 8D7 monoclonal antibody recognizes Asialoglycoprotein receptor 1 (ASGPR 1), also known as Hepatic lectin H1 (HL-1). ASGPR 1 is an approximately 42 kDA type II integral membrane protein that is expressed on the surface of hepatic cells. It is expressed by hepatocytes on the sinusoidal-lateral plasma membrane but not on the bile canalicular membrane. ASGPR 1 plays a role in serum glycoprotein homeostasis. It functions as a subunit of the Asialoglycoprotein receptor (ASGPR) complex that binds, internalizes, and transports various glycoproteins for lysosomal degradation. The receptor may also promote hepatic infection by the binding and uptake of various viruses. The immunogen used to generate the 8D7 hybridoma was rat liver membrane extracts. Rat ASGPR consists of three polypeptide subunits (Rat hepatic lectin 1-3 (RHL1-3). The 8D7 antibody has been shown to react with a subunit-specific epitope on RHL-1. Clone 8D7 cross-reacts with human ASGPR 1.

Specifications
ASGPR 1 | |
Monoclonal | |
0.5mg/mL | |
Asialoglycoprotein receptor 1, ASGR1, ASGP-R1, ASGPR 1, HL-1, CLEC4H1 | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Rat | |
IgG1 κ |
Flow Cytometry | |
8D7 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat Liver membrane constituents | |
100 μg | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.