Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

aspartoacylase (Canavan disease), Mouse, Clone: 3C11, Abnova™

Catalog No. 89014057 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-057 Supplier Abnova Corporation Supplier No. H00000443M09
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ASPA.

This gene encodes an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues. Mutations in this gene cause Canavan disease. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq

Sequence: MTSCHIAEEHIQKVAIFGGTHGNELTGVFLVKHWLENGAEIQRTGLEVKPFITNPRAVKKCTRYIDCDLNRIFDLENLGKKMSEDLPYEVRRAQEINHLF

Specifications

Antigen aspartoacylase (Canavan disease)
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3C11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ASPA.
Formulation PBS with no preservative; pH 7.4
Gene ASPA
Gene Accession No. NM_000049
Gene Alias ACY2/ASP
Gene Symbols ASPA
Host Species Mouse
Immunogen ASPA (NP_000040, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 443
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.