Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alveolar soft part sarcoma chromosome region, candidate 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018744 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-744 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-744 Supplier Abnova Corporation Supplier No. H00079058D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human ASPSCR1 protein.

This gene is a candidate gene for alveolar soft part sarcoma (ASPS). It has been found that this gene is fused with transcription factor TFE3 gene in ASPS and also in renal cell carcinomas. Several alternatively spliced transcript variants of this gene have been described, but their full length nature has not been determined. [provided by RefSeq

Sequence: MAAPAGGGGSAVSVLAPNGRRHTVKVTPSTVLLQVLEDTCRRQDFNPCEYDLKFQRSVLDLSLQWRFANLPNNAKLEMVPASRSREGPENMVRIALQLDDGSRLQDSFCSGQTLWELLSHFPQIRECLQHPGGATPVCVYTRDEVTGEAALRGTTLQSLGLTGGSATIRFVMKCYDPVGKTPGSLGSSASAGQAAASAPLPLESGELSRGDLSRPEDADTSGPCCEHTQEKQSTRAPAAAPFVPFSGGGQRQGGPPGPTRPLTSSSAKLPKSLSSPGGPSKPKKSKSGQDPQQEQEQERERDPQQEQERERPVDREPVDREPVVCHPDLEERLQAWPAELPDEFFELTVDDVRRRLAQLKSERKRLEEAPLVTKAFREAQIKEKLERYPKVALRVLFPDRYVLQGFFRPSETVGDLRDFVRSHLGNPELSFYLFITPPKTVLDDHTQTLFQANLFPAALVHLGAEEPAGVYLEPGLLEHAISPSAADVLVARYMSRAAGSPSPLPAPDPAPKSEPAAEEGALVPPEPIPGTAQPVKRSLGKVPKWLKLPASKR

Specifications

Antigen alveolar soft part sarcoma chromosome region, candidate 1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ASPSCR1 protein.
Formulation PBS with no preservative; pH 7.4
Gene ASPSCR1
Gene Accession No. BC018722.1
Gene Alias ASPCR1/ASPL/ASPS/RCC17/TUG/UBXD9/UBXN9
Gene Symbols ASPSCR1
Host Species Rabbit
Immunogen ASPSCR1 (AAH18722.1, 1 a.a. ∼ 553 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79058
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.