Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASS1 Mouse anti-Human, Rat, Mouse, Monkey, Clone: 2C10, MilliporeSigma™

Catalog No. MABN704MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN704MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN704MI Supplier MilliporeSigma Supplier No. MABN704
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Argininosuccinate synthase using this mouse monoclonal antibody, Anti-ASS1 Antibody, clone 2C10 validated for use in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASS1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2C10
Conjugate Unconjugated
Formulation Purified mouse monoclonal in PBS with up to 0.1% sodium azide and 0.5% protein stabilizer.
Gene Symbols ASS, CTLN1, ASS1
Host Species Mouse
Immunogen Purified recombinant fragment of human ASS1 expressed in E. Coli.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Monkey, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.