Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Astrocytomas Mouse anti-Guinea Pig, Human, Mouse, Rat, Clone: J1-31, MilliporeSigma™

Catalog No. MAB5372MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5372MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5372MI Supplier MilliporeSigma Supplier No. MAB5372
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Astrocytomas using this Anti-Astrocytomas Antibody, a marker for Low Grade Astrocytoma, clone J1-31 validated for use in western blot, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Astrocytomas
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone J1-31
Conjugate Unconjugated
Formulation Ascites fluid. Liquid.
Host Species Mouse
Immunogen Human cerebral white matter plaque materials from a multiple sclerosis patient.
Quantity 100 μL
Primary or Secondary Primary
Target Species Guinea Pig, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.