Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AT2 Receptor Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB15554200U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15554200U 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15554200U Supplier MilliporeSigma Supplier No. AB15554200UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect AT2 Receptor using this Anti-AT2 Receptor Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AT2 Receptor
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.025% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).
Gene Symbols AGTR2; MRX88; AT2; ATGR2
Host Species Rabbit
Immunogen Purified peptide from the N-terminus of rat AT2 Receptor (Accession number P35351).
Purification Method Immunoaffinity purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000686.4
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.