Learn More
Description
Specifications
Specifications
| Antigen | AT2 Receptor |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.025% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min). |
| Gene Symbols | AGTR2; MRX88; AT2; ATGR2 |
| Host Species | Rabbit |
| Immunogen | Purified peptide from the N-terminus of rat AT2 Receptor (Accession number P35351). |
| Purification Method | Affinity Purified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.