Learn More
ATAD3B, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ATAD3B peptide using ARM Technology.
Supplier: Abnova Corporation H00083858K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
ATAD3A (MIM 612316) and ATAD3B are mitochondrial membrane proteins that contribute to the stabilization of large mitochondrial DNA (mtDNA)-protein complexes called nucleoids (He et al., 2007 [PubMed 17210950]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ATAD3B | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ATAD3B | |
ATAD3B | |
A synthetic peptide of human ATAD3B | |
RUO | |
83858 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ATPase family, AAA domain containing 3B | |
ATAD3B | |
AAA-TOB3, KIAA1273, TOB3 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.