Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ataxin-7 Mouse anti-Rat, Mouse, Human, Clone: 3SCA-1C1, MilliporeSigma™

Catalog No. 041573MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-157-3MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-157-3MI Supplier MilliporeSigma Supplier No. 041573
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Ataxin-7 Antibody, clone 3SCA-1C1 is an antibody against Ataxin-7 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ataxin-7
Applications Western Blot
Classification Monoclonal
Clone 3SCA-1C1
Conjugate Unconjugated
Formulation Unpurified mouse IgG1, k antibody in ascites fluid without preservatives.
Gene Accession No. NP_000324
Gene Symbols ATXN7
Host Species Mouse
Immunogen Recombinant human Ataxin 7 as a fusion protein (aa 1 to aa 229).
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.