Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

arginyltransferase 1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004534 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-534 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-534 Supplier Abnova Corporation Supplier No. H00011101A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ATE1.

This gene encodes an arginyltransferase, an enzyme that is involved in posttranslational conjugation of arginine to N-terminal aspartate or glutamate residues. Conjugation of arginine to the N-terminal aspartate or glutamate targets proteins for ubiquitin-dependent degradation. Alternative splicing results in two transcript variants encoding distinct isoforms. [provided by RefSeq

Sequence: MAFWAGGSPSVVDYFPSEDFYRCGYCKNESGSRSNGMWAHSMTVQDYQDLIDRGWRRSGKYVYKPVMNQTCCPQYTIRCRPLQFQPS

Specifications

Antigen arginyltransferase 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ATE1.
Formulation 50% glycerol
Gene ATE1
Gene Accession No. NM_001001976
Gene Alias MGC26724
Gene Symbols ATE1
Host Species Mouse
Immunogen ATE1 (NP_001001976, 1 a.a. ∼ 87 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 11101
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.