Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

activating transcription factor 4 (tax-responsive enhancer element B67), Mouse, Clone: 2B3, Abnova™

Catalog No. 89014058 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-058 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-058 Supplier Abnova Corporation Supplier No. H00000468M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ATF4.

This gene encodes a transcription factor that was originally identified as a widely expressed mammalian DNA binding protein that could bind a tax-responsive enhancer element in the LTR of HTLV-1. The encoded protein was also isolated and characterized as the cAMP-response element binding protein 2 (CREB-2). The protein encoded by this gene belongs to a family of DNA-binding proteins that includes the AP-1 family of transcription factors, cAMP-response element binding proteins (CREBs) and CREB-like proteins. These transcription factors share a leucine zipper region that is involved in protein-protein interactions, located C-terminal to a stretch of basic amino acids that functions as a DNA binding domain. Two alternative transcripts encoding the same protein have been described. Two pseudogenes are located on the X chromsome at q28 in a region containing a large inverted duplication. [provided by RefSeq

Sequence: SSTPDHSFSLELGSEVDITEGDRKPDYTAYVAMIPQCIKEEDTPSDNDSGICMSPESYLGSPQHSPSTRGSPNRSLPSPGVLCGSARPKPYDPPGEKMVA

Specifications

Antigen activating transcription factor 4 (tax-responsive enhancer element B67)
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ATF4.
Formulation PBS with no preservative; pH 7.4
Gene ATF4
Gene Accession No. NM_001675
Gene Alias CREB-2/CREB2/TAXREB67/TXREB
Gene Symbols ATF4
Host Species Mouse
Immunogen ATF4 (NP_001666.2, 171 a.a. ∼ 270 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 468
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.