Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATF6 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 09069MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-069-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-069-MI Supplier MilliporeSigma Supplier No. 09069
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect ATF6 using this Anti-ATF6 Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified rabbit polyclonal IgG in storage buffer containing PBS with 0.05% sodium azide.
Gene Symbols ATF6; activating transcription factor 6
Host Species Rabbit
Immunogen KLH-tagged synthestic peptide synthesized to the N-terminal region of human ATF6 designated as the Cytoplasmic Domain.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_031374
Target Species Human
Content And Storage −20°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.