Learn More
Description
Autophagy is a process of bulk degradation of cytoplasmic components by the lysosome or vacuole. Human ATG3 displays the same enzymatic characteristics in vitro as yeast Apg3, a protein-conjugating enzyme essential for autophagy (Tanida et al., 2002 [PubMed 11825910]).[supplied by OMIM
Specifications
Specifications
| Antigen | ATG3 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1G3 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant ATG3. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ATG3 |
| Gene Accession No. | NM_022488 |
| Gene Alias | APG3/APG3-LIKE/APG3L/DKFZp564M1178/FLJ22125/MGC15201/PC3-96 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.