Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATG7 Rabbit Monoclonal Rabbit anti-Human, Clone: EPR6251, MilliporeSigma™

Catalog No. MABN1124MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1124MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1124MI Supplier MilliporeSigma Supplier No. MABN1124

Rabbit Monoclonal Antibody

This Anti-ATG7, clone EPR6251, Rabbit Monoclonal Antibody is validated for use in Western Blotting and Immunofluorescence for the detection of ATG7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATG7
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone EPR6251
Conjugate Unconjugated
Formulation Rabbit Monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human ATG7.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) ATG7, APG7L
Target Species Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.