Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATL3 Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524652
Description
This antibody is predicted to react with rat based on sequence homology.
ATL3 is GTPase tethering membranes through formation of trans-homooligomer and mediating homotypic fusion of endoplasmic reticulum membranes. ATL3 is function in endoplasmic reticulum tubular network biogenesis.
Specifications
ATL3 | |
Polyclonal | |
ATL3 | |
atlastin 3, atlastin-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100752365, 109168, 25923 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q6DD88, Q91YH5 | |
ATL3 | |
KLH conjugated synthetic peptide between 217-245 amino acids from the central region of human ATL3 | |
400 μL | |
Primary | |
Hamster, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction