Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATM/Chk2 Pathway Explorer Mouse anti-Rat, Mouse, Rat, Clone: AM9; 10H11.E12; 7, MilliporeSigma™

Catalog No. 15116MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-116-MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-116-MI Supplier MilliporeSigma Supplier No. 15116
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Antibody pack contains Anti-ATM Antibody, Anti-phospho-ATM Antibody (Ser1981), Anti-Chk2 Antibody validated for use in western blot, immunocytochemistry and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATM/Chk2 Pathway Explorer Antibody MiniPack
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation 3 individual tubes containing either Anti-ATM, clone AM9; Anti-phospho-ATM (Ser1981), clone 10H11.E12; or Anti-Chk2, clone 7
Gene Accession No. NP_000042; NP_001005735
Gene Symbols AT1, ATA, ATC, ATD, ATDC, ATE, DKFZp781A0353, EC 2.7.11.1, MGC74674, TEL1, TELO1, CHETK-alpha , CHK , CK , CKI, EC 2.7.1.32
Host Species Mouse
Immunogen 05-740SP: KLH-conjugated, synthetic peptide corresponding to amino acids 1974-1988 (SLAFEEG[pS]QSTTISS) of human ATM, synthetic peptide corresponding to amino acids 1974-1988 (SLAFEEG[pS]QSTTISS) of human ATM. The immunizing sequence has 11/12
Quantity 1 Unit
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage −20°C for up to 12 months.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.