Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATM, Mouse, Clone: 2C10, Abnova™

Catalog No. 89123139 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-139 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-139 Supplier Abnova Corporation Supplier No. H00000472M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ATM.

The protein encoded by this gene belongs to the PI3/PI4-kinase family. This protein is an important cell cycle checkpoint kinase that phosphorylates; thus, it functions as a regulator of a wide variety of downstream proteins, including tumor suppressor proteins p53 and BRCA1, checkpoint kinase CHK2, checkpoint proteins RAD17 and RAD9, and DNA repair protein NBS1. This protein and the closely related kinase ATR are thought to be master controllers of cell cycle checkpoint signaling pathways that are required for cell response to DNA damage and for genome stability. Mutations in this gene are associated with ataxia telangiectasia, an autosomal recessive disorder. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MTLHEPANSSASQSTDLCDFSGDLDPAPNPPHFPSHVVKATFAYISNCHKTKLKSILEILSKSPDSYQKILLAICEQAAETNNVYKKHRILKIYHLFVSLLLKDIKSGLGGAWAFVLRDVIYTLIHYINQRKLTIFSQ

Specifications

Antigen ATM
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 2C10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant ATM.
Formulation PBS with no preservative; pH 7.4
Gene ATM
Gene Accession No. BC007023.1
Gene Alias AT1/ATA/ATC/ATD/ATDC/ATE/DKFZp781A0353/MGC74674/TEL1/TELO1
Gene Symbols ATM
Host Species Mouse
Immunogen ATM (AAH07023.1, 1 a.a. ∼ 138 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 472
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.