Learn More
Description
Specifications
Specifications
| Antigen | ATM |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | AM9 |
| Conjugate | Unconjugated |
| Formulation | mouse ascites containing 0.05% sodium azide |
| Gene Accession No. | NM_000051 and others |
| Gene Symbols | ATM, AT, TELO1, ATC, DKFZp781A0353, MGC74674, ATD, ATA, AT1, T-PLL, ATE, TEL1, ATDC, TPLL |
| Host Species | Mouse |
| Immunogen | Full-length human ATM |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.