Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATM Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 071286MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-128-6MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-128-6MI Supplier MilliporeSigma Supplier No. 071286
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-ATM Antibody detects level of ATM and has been published and validated for use in western blotting and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATM
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols AT1; ATA; ATC; ATD; ATDC; ATE; DKFZp781A0353 2; EC 2.7.11.1 3; MGC74674; TEL1; TELO1
Host Species Rabbit
Immunogen KLH-conjugated synthetic peptide corresponding to sequence derived from the Abl-interacting domain of human ATM.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_000042.
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.