Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATOH1, Mouse, Clone: 3B12, Abnova™

Catalog No. 89016242 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-242 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-242 Supplier Abnova Corporation Supplier No. H00000474M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ATOH1.

This protein belongs to the basic helix-loop-helix (BHLH) family of transcription factors. It activates E-box dependent transcription along with E47. [provided by RefSeq

Sequence: PTPPGSCRTRFSAPASAGGYSVQLDALHFSTFEDSALTAMMAQKNLSPSLPGSILQPVQEENSKTSPRSHRSDGEFSPHSHYSDSDEAS

Specifications

Antigen ATOH1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3B12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ATOH1.
Formulation PBS with no preservative; pH 7.4
Gene ATOH1
Gene Accession No. NM_005172
Gene Alias ATH1/HATH1/MATH-1/bHLHa14
Gene Symbols ATOH1
Host Species Mouse
Immunogen ATOH1 (NP_005163.1, 266 a.a. ∼ 354 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 474
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.