Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATOH1, Mouse, Clone: 4D10, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ATOH1.
Supplier: Abnova Corporation H00000474M10
Description
This protein belongs to the basic helix-loop-helix (BHLH) family of transcription factors. It activates E-box dependent transcription along with E47. [provided by RefSeq
Sequence: MSRLLHAEEWAEVKELGDHHRQPQPHHLPQPPPPPQPPATLQAREHPVYPPELSLLDSTDPRAWLAPTLQGICTARAAQYLLHSPELGASEAAAPRDEVDGRGELVRRSSGGASSSKSPGPVKVREQLCKSpecifications
ATOH1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005172 | |
ATOH1 | |
ATOH1 (NP_005163, 1 a.a. ∼ 130 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
4D10 | |
Mouse monoclonal antibody raised against a full length recombinant ATOH1. | |
ATOH1 | |
ATH1/HATH1/MATH-1/bHLHa14 | |
Mouse | |
Affinity chromatography | |
RUO | |
474 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction