Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP Synthase Mouse anti-Human, Mouse, Rat, Clone: 4.3E8.D1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3494
Description
Anti-ATP Synthase Antibody, β chain, clone 4.3E8.D1 detects level of ATP Synthase and has been published and validated for use in IC and immunoprecipitation.
Specifications
ATP Synthase | |
Monoclonal | |
Unconjugated | |
NM_001686.3 | |
Mouse | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Immunocytochemistry, Immunoprecipitation | |
4.3E8.D1 | |
Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl with 0.1% sodium azide. | |
ATP5B, ATPMB, ATPSB, MGC5231, EC 3.6.3.14 | |
Rat brain mitochondria | |
Primary | |
2-8°C in undiluted aliquots for up to 6 months. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction