Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-ATP Synthase subunit beta Clone: 11/21-7-A8,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABS1304
Description
Specifically detects ATP Synthase subunit β Clone: 11/21-7-A8 in Human, Mouse, Rat samples, and it is validated for Dot Blot, ELISA, Immunocytochemistry, Western Blotting
ATP synthase subunit beta, mitochondrial (EC 3.6.3.14; UniProt P06576; also known as ATP synthase H+ transporting mitochondrial F1 complex beta polypeptide, beta-F1-ATPase, Epididymis secretory protein Li 271, Mitochondrial ATP synthase beta subunit, Mitochondrial ATP synthetase beta subunit) is encoded by the ATP5B (also known as ATPMB, ATPSB, HEL-S-271) gene (Gene ID 506) in human. Mitochondrial ATP synthase produces ATP from ADP in the presence of a proton gradient generated by electron transport complexes. This ATPase contains two structural domains, F1-containing extramembrane catalytic core, and F0-containing the membrane proton channel that are linked via a central stalk and a peripheral stalk. Subunits alpha and beta form the catalytic code in F1. Tumor development requires the selection of cancer cells with a repressed biogenesis and functional activity of mitochondria. Both beta-F1-ATPase/GAPDH and beta-F1-ATPase/Hsp60 ratios are found to be significantly lower in tumors than the corresponding normal tissues. Studies conducted in human colon cancer cell line HCT116 show the involvement of AMPK (AMP-activated protein kinase) and GCN2 (general control non-derepressible 2; eIF2α kinase) in the onset of colon cancer progression by repressing of beta-F1-ATPase synthesis and promoting the abnormal bioenergetics of mitochondria.
Specifications
| ATP Synthase subunit β | |
| Monoclonal | |
| Unconjugated | |
| P06576 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Dot Blot, ELISA, Immunocytochemistry, Western Blot | |
| 11/21-7-A8 | |
| Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| ATP5B, ATPMB, ATPSB | |
| His-tagged recombinant protein corresponding to human ATP Synthase subunit β. | |
| 100 μg | |
| Signaling | |
| NP_001677 | |
| 2°C to 8°C for one year from date of shipment | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction