Learn More
ATP13A2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ATP13A2.
Supplier: Abnova Corporation PAB12479
Description
This gene encodes a member of the P5 subfamily of ATPases which transports inorganic cations as well as other substrates. Mutations in this gene are associated with Kufor-Rakeb syndrome (KRS), also referred to as Parkinson disease 9. Multiple transcript variants encoding different isoforms have been found for this gene
Specifications
ATP13A2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ATP13A2. | |
In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
Q9NQ11 | |
ATP13A2 | |
A synthetic peptide corresponding to amino acids 200-300 of human ATP13A2. | |
RUO | |
23400 | |
Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (2 ug/mL) Immunohistochemistry (10-20 ug/mL) The optimal working dilution should be determined by the end user. | |
ATP13A2 | |
FLJ26510/HSA9947/KRPPD/PARK9 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only