Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP13A2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003135 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-135 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-135 Supplier Abnova Corporation Supplier No. H00023400A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ATP13A2.

This gene encodes a member of the P5 subfamily of ATPases which transports inorganic cations as well as other substrates. Mutations in this gene are associated with Kufor-Rakeb syndrome (KRS), also referred to as Parkinson disease 9. Multiple transcript variants encoding different isoforms have been found for this gene

Sequence: KPLWGVRLRLRPCNLAHAETLVIEIRDKEDSSWQLFTVQVQTEAIGEGSLEPSPQSQAEDGRSQAAVGAVPEGAWKDTAQLHKSEEA

Specifications

Antigen ATP13A2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ATP13A2.
Formulation 50% glycerol
Gene ATP13A2
Gene Accession No. NM_022089
Gene Alias FLJ26510/HSA9947/KRPPD/PARK9
Gene Symbols ATP13A2
Host Species Mouse
Immunogen ATP13A2 (NP_071372, 68 a.a. ∼ 154 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23400
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.