Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP13A3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525831
Description

Specifications
ATP13A3 | |
Polyclonal | |
ATP13A3 | |
AFURS1, ATPase family homolog up-regulated in senescence cells 1, probable cation-transporting ATPase 13A3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79572 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9H7F0 | |
ATP13A3 | |
KLH conjugated synthetic peptide between 101-129 amino acids from the N-terminal region of human ATP13A3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction