Learn More
Description
This gene encodes one of the SERCA Ca(2+)-ATPases, which are intracellular pumps located in the sarcoplasmic or endoplasmic reticula of muscle cells. This enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen, and is involved in muscular excitation and contraction. Mutations in this gene cause some autosomal recessive forms of Brody disease, characterized by increasing impairment of muscular relaxation during exercise. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq]
Specifications
Specifications
| Antigen | ATP2A1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2C9 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant ATP2A1. |
| Formulation | ascites with no preservative |
| Gene | ATP2A1 |
| Gene Accession No. | NM_173201 |
| Gene Alias | ATP2A/SERCA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.