Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP5F1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP133465
Description
ATP5F1 Polyclonal specifically detects ATP5F1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ATP5F1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
ATP5PB | |
Recombinant protein encompassing a sequence within the center region of human ATP synthase B1. The exact sequence is proprietary. | |
0.1 mL | |
metabolism | |
515 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.45mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
ATP synthase B chain, mitochondrial, ATP synthase subunit b, mitochondrial, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit b, isoform 1, ATP synthase, H+ transporting, mitochondrial F0 complex, subunit B1, ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1, ATPase subunit b, cell proliferation-inducing protein 47, H+-ATP synthase subunit b, MGC24431, PIG47 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction