Learn More
ATP6V1B1, Mouse, Clone: 3G11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ATP6V1B1.
Supplier: Abnova Corporation H00000525M02
Description
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c″, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is one of two V1 domain B subunit isoforms and is found in the kidney. Mutations in this gene cause distal renal tubular acidosis associated with sensorineural deafness. [provided by RefSeq]
Sequence: MAMEIDSRPGGLPGSSCNLGAAREHMQAVTRNYITHPRVTYRTVCSVNGPLVVLDRVKFAQYAEIVHFTLPDGTQSpecifications
ATP6V1B1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001692 | |
ATP6V1B1 | |
ATP6V1B1 (NP_001683.2, 1 a.a. ∼ 75 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA | |
3G11 | |
Mouse monoclonal antibody raised against a partial recombinant ATP6V1B1. | |
ATP6V1B1 | |
ATP6B1/MGC32642/RTA1B/VATB/VMA2/VPP3 | |
Mouse | |
Affinity chromatography | |
RUO | |
525 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.