Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1G2, Mouse, Clone: 2E11, Abnova™

Catalog No. 89016283 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-283 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-283 Supplier Abnova Corporation Supplier No. H00000534M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ATP6V1G2.

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c″, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is one of three V1 domain G subunit proteins. This gene had previous gene symbols of ATP6G and ATP6G2. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq]

Sequence: QMEVEQYRREREHEFQSKQQAAMGSQGNLSAEVEQATRRQVQGMQSSQQRNRERVLAQLLGMVCDVRPQVHPNYRISA

Specifications

Antigen ATP6V1G2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ATP6V1G2.
Formulation PBS with no preservative; pH 7.4
Gene ATP6V1G2
Gene Accession No. NM_130463
Gene Alias ATP6G/ATP6G2/NG38/VMA10
Gene Symbols ATP6V1G2
Host Species Mouse
Immunogen ATP6V1G2 (NP_569730, 41 a.a. ∼ 118 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 534
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.