Learn More
ATP7A, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA120840
Description
Copper-transporting ATPase 1 is an integral membrane protein cycling constitutively between the trans-golgi network and the plasma membrane. It may supply copper to copper-requiring proteins within the secretory pathway, when localized in the trans-golgi network. Under conditions of elevated extracellular copper, it relocalized to the plasma membrane where it functions in the efflux of copper from cells.
PA1-20840 detects ATP7A in human, mouse and rat samples.PA1-20840 has been successfully used in Western blot procedures.The PA1-20840 immunogen is a recombinant fragment, corresponding to amino acids 1407-1500 of Human ATP7A..
Specifications
ATP7A | |
Polyclonal | |
Unconjugated | |
Q04656, Q64430, P70705 | |
ATP7A | |
Recombinant fragment, corresponding to amino acids 1407-1500 of Human ATP7A. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Western Blot | |
1mg/mL | |
ATP7A | |
RP3-465G10.1, FLJ17790, MK, MNK | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
538 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.