Learn More
ATR, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA123182
Description
Ataxia Telangiectasia Mutated (ATM) and Rad3-related (ATR) protein is a member of the phosphatidyl-inositol 3-kinase (PI 3-kinase) like family of protein kinases. ATR has a predicted molecular mass of approximately 301 kDa, and it has been shown to be involved in cellular responses to DNA damage. Specifically, it plays a role in checkpoint activation in response to either the presence of stalled replication forks, or by the presence of lesions caused by UV light. Inactivation of ATR is an embryonic lethal event in mice.
PA1-23182 detects ATR in human, mouse and hamster samples.PA1-23182 has been successfully used in Western Blot procedures.PA1-23182 immunogen corresponds to The carboxy-terminal portion of human ATR protein (amino acids 2122-2644).Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Specifications
ATR | |
Polyclonal | |
ATR | |
FRP1, MEC1, SCKL, SCKL1 | |
Rabbit | |
50 μL | |
Primary | |
Hamster, Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Q13535, Q9JKK8 | |
Atr | |
The carboxy-terminal portion of human ATR protein (amino acids 2122-2644). | |
RUO | |
545 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.