Learn More
Attractin Mouse anti-Human, Unlabeled, Clone: 14, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 611530
Description
Interactions between cell adhesion molecules and extracellular matrix (ECM) components mediate the migration of antigen specific T cells from the bloodstream to inflammatory sites. CD26, a surface marker of these T cells, is identical to dipeptidyl peptidase (DPPIV). Attractin was identified in serum as a soluble 175kDa form of DPPIV, termed DPPT-L. It is a member of the CUB domain familly, which includes the complement proteins C1r/C1s, Uegf, and BMP-1 (procollagen C proteinase), a protein that regulates cell interactions during development.
Although unable to induce cell proliferation on its own, attractin enhances the proliferative response of PBMC in the presence of antigen. Attractin mediates the spreading of monocytes (antigen presenting cells; APCs) which become the focus for T cell clustering and formation of the immunoregulatory unit. Thus, attractin is thought to modulate the interaction between T cells and APCs, thereby enhancing the effectiveness of antigen presentation. In addition, attractin is the human homolog of the mahogany protein, a transmembrane signaling receptor
involved in suppression of the obesity observed in mice with the agouti-lethal-yellow mutation.
Host Species: Mouse
Clone: 14
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Attractin aa. 184-383
Formula Weight [Chemical]: 175kDa
Immunofluorescence, Western Blotting
Specifications
| Attractin | |
| Monoclonal | |
| 250μg/mL | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| IgG1 |
| Immunofluorescence, Western Blot | |
| 14 | |
| Unconjugated | |
| Human Attractin aa. 184-383 | |
| 50 μg | |
| Cell Biology | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.