Learn More
AUH, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human AUH peptide using ARM Technology.
Supplier: Abnova Corporation H00000549K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
AU-specific RNA-binding enoyl-CoA hydratase (AUH) protein binds to the AU-rich element (ARE), a common element found in the 3' UTR of rapidly decaying mRNA such as c-fos, c-myc and granulocyte/ macrophage colony stimulating factor. ARE elements are involved in directing RNA to rapid degradation and deadenylation. AUH is also homologous to enol-CoA hydratase, an enzyme involved in fatty acid degradation, and has been shown to have intrinsic hydratase enzymatic activity. AUH is thus a bifunctional chimera between RNA binding and metabolic enzyme activity. A possible subcellular localization in the mitochondria has been demonstrated for the mouse homolog of this protein which shares 92% identity with the human protein. It has been suggested that AUH may have a novel role as a mitochondrial located AU-binding protein. Human AUH is expressed as a single mRNA species of 1.8kb, and translated as a 40kDa precursor protein which is subsequently processed to a 32kDa mature form. [provided by RefSeq]
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| AUH | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| AUH | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 1mg | |
| AU RNA binding protein/enoyl-Coenzyme A hydratase | |
| AUH | |
| AUH | |
| A synthetic peptide of human AUH | |
| RUO | |
| 549 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.