Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aurora B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514076
Description
Chromosomal segregation during mitosis as well as meiosis is regulated by kinases and phosphatases. The Aurora kinases associate with microtubules during chromosome movement and segregation. STK12 (Aurora kinase B) localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers, and Aurora kinase A localizes to centrosomes
Specifications
Aurora B | |
Polyclonal | |
AURKB | |
Serine/threonine-protein kinase 12; Aurora-B; Aurora- and Ipl1-like midbody-associated protein 1; AIM-1; Aurora/IPL1-related kinase 2; Aurora-related kinase 2; STK-1; AURKB; AIK2; AIM1; ARK2; STK12,NP_004208; Q96GD4 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
20877, 9212 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q96GD4, O70126 | |
AURKB | |
KLH conjugated synthetic peptide between 283-313 amino acids from the central region of human Aurora-B (ARK/STK12) | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction