Learn More
Description
Specifications
Specifications
| Antigen | Avian Influenza A H6N1 Nucleoprotein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-2 ug/ml |
| Formulation | PBS containing 0.05% BSA with 0.05% Sodium Azide |
| Gene Alias | NP, Nucleoprotein |
| Host Species | Rabbit |
| Immunogen | Amino acids 58-77 of avian flu nucleoprotein (accession number Q71FW9) were used as the immunogen. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
