Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AVIL Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524690
Description
This antibody is predicted to react with mouse based on sequence homology.
AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.
Specifications
| AVIL | |
| Polyclonal | |
| AVIL | |
| ADVIL, DOC6, p92 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10677 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| O75366 | |
| AVIL | |
| KLH conjugated synthetic peptide between 176-204 amino acids from the N-terminal region of human AVIL | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction