Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AWAT2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524313
Description
This gene encodes an enzyme belonging to the diacylglycerol acyltransferase family. This enzyme produces wax esters by the esterification of long chain (or wax) alcohols with acyl-CoA-derived fatty acids. It functions in lipid metabolism in the skin, mostly in undifferentiated peripheral sebocytes. This enzyme may also have acyl-CoA:retinol acyltransferase activities, where it catalyzes the synthesis of diacylglycerols and retinyl esters.
Specifications
AWAT2 | |
Polyclonal | |
AWAT2 | |
DC4, DGAT2L4, MFAT, WS, diacylglycerol O-acyltransferase 2-like protein 4, diacylglycerol O-acyltransferase candidate 4, long-chain-alcohol O-fatty-acyltransferase 2, multifunctional O-acyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
158835, 245532 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q6E213, Q6E1M8 | |
AWAT2 | |
KLH conjugated synthetic peptide between 303-331 amino acids from the C-terminal region of human AWAT2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction