Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axin-1 Mouse anti-Human, Mouse, Clone: A5, MilliporeSigma™

Catalog No. 051579MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-157-9MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-157-9MI Supplier MilliporeSigma Supplier No. 051579
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Axin-1 using this Anti-Axin-1 Antibody, clone A5 validated for use in western blot, immunoprecipitation and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Axin-1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A5
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_033863.2
Gene Symbols AXIN, hAxin
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the N-terminus of mouse Axin-1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.