Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human B cells (CD19) Mouse anti-Human, Clone: FMC63, MilliporeSigma™

Catalog No. MAB1794MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1794MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1794MI Supplier MilliporeSigma Supplier No. MAB1794
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Human B cells CD19 Antibody, clone FMC63 is validated for use in flow cytometry, immunofluorescence for the detection of CD19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B cells (CD19)
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone FMC63
Conjugate Unconjugated
Formulation Purified from mouse ascitic fluid. The antibody is supplied in 25mM Tris-HCl buffer, 0.4MNaCl, pH 8.0, containing 0.2% bovine serum albumin and 0.1% sodium azide. The characteristics of each lot are tested by electrophoresis and flow cytometry.
Gene Accession No. NM_001770.4
Gene Symbols CD19, B4, MGC12802, Leu-12
Host Species Mouse
Immunogen Raji cells
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.