Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-Myb Mouse anti-Human, Mouse, Clone: LX015.1, MilliporeSigma™

Catalog No. MABE886MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE886MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE886MI Supplier MilliporeSigma Supplier No. MABE886
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect B-Myb using this Anti-B-Myb Antibody, clone LX015.1 validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B-Myb
Applications Western Blot
Classification Monoclonal
Clone LX015.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002457
Gene Symbols MYBL2, BMYB
Host Species Mouse
Immunogen Recombinant protein corresponding to human B-Myb.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.