Learn More
B2 Bradykinin Receptor Mouse, Unlabeled, Clone: 20, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610452
Description
Bradykinin is a nine amino acid vasoactive peptide that elicits numerous physiologic responses such as vasodilation, smooth muscle spasm, and pain. Bradykinin is one of a family of such peptides called the kinins. The kinins are generated from high molecular weight precursors called kininogens from proteolysis induced by pathophysiologic conditions such as inflammation or allergy. The physiological actions of these kinins are mediated by their interaction with transmembrane receptors. There are two distinct bradykinin receptor subtypes: B1 and B2. Both are coupled to G-proteins. The B2 receptor subtype is found in healthy smooth muscle cells and neurons, whereas, the B1 receptors are only detected following tissue injury. The B2 receptor is similar in structure to other seven helix G-protein coupled receptors. Bradykinin has a relatively low affinity for B1 receptors, and interacts primarily with the B2 receptor. This interaction stimulates several second messenger systems, including inositol phospholipid hydrolysis, arachidonic acid metabolism, tyrosine phosphorylation, and membrane depolarization and hyperpolarization.

Specifications
B2 Bradykinin Receptor | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
150 μg | |
Cell Biology | |
Human, Rat | |
IgG2b κ |
Western Blot | |
20 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.